2-Bromoaniline hydrochloride ≥97.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAB0870-25GEA 1275 CNY
TCIAB0870-25G
2-Bromoaniline hydrochloride ≥97.0% (通过HPLC,总氮)
2-Bromoaniline hydrochloride
分子式: C₆H₆BrN
分子量: 208,48 g/mol
MDL编号: MFCD00060215
CAS号: 94718-79-3

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR