3-Benzalbutyramide ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB0892-5GEA 0 CNY
TCIAB0892-5G
3-Benzalbutyramide ≥98.0% (通过GC)
3-Benzalbutyramide
分子式: C₁₁H₁₃NO
分子量: 175,23 g/mol
MDL编号: MFCD00059860
CAS号: 7236-47-7
EINECS: 230-637-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 132.0 to 135.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR