2-(Butylthio)ethanol ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB0917-5MLEA 0 CNY
TCIAB0917-5ML
2-(Butylthio)ethanol ≥98.0% (通过GC)
2-(Butylthio)ethanol
分子式: C₆H₁₄OS
分子量: 134,24 g/mol
沸点: 217°C
MDL编号: MFCD00014042
CAS号: 5331-37-3
EINECS: 226-229-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR