4-tert-Butylphenyl acetate ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAB0962-5MLEA 0 CNY
TCIAB0962-5ML
4-tert-Butylphenyl acetate ≥97.0% (通过GC)
4-tert-Butylphenyl acetate
分子式: C₁₂H₁₆O₂
分子量: 192,26 g/mol
沸点: 245°C
MDL编号: MFCD00026308
CAS号: 3056-64-2
EINECS: 221-288-6

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR