2-Isopropoxyethyl benzoate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAB0972-5MLEA 0 CNY
TCIAB0972-5ML
2-Isopropoxyethyl benzoate ≥95.0% (通过GC)
2-Isopropoxyethyl benzoate
分子式: C₁₂H₁₆O₃
分子量: 208,26 g/mol
MDL编号: MFCD00059357
CAS号: 95241-36-4

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR