2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium chloride hydrate ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAB1047-100MGEA 0 CNY
TCIAB1047-100MG
2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium chloride hydrate ≥98.0% (通过滴定分析)
2,3-Bis(4-nitrophenyl)-5-phenyltetrazolium chloride hydrate
分子式: C₁₉H₁₃ClN₆O₄
MDL编号: MFCD00060010
CAS号: 69231-13-6
UN号: 1325
ADR: 4.1,III

现在订购

产品技术规格

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR