2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium chloride ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAB1054-100MGEA 0 CNY
TCIAB1054-100MG
2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium chloride ≥98.0% (通过HPLC)
2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium chloride
分子式: C₁₉H₁₃ClF₂N₄
分子量: 370,79 g/mol
MDL编号: MFCD00059998
CAS号: 240800-46-8

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR