2,3-Bis(4-methoxyphenyl)-5-phenyltetrazolium chloride ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAB1056-1GEA 0 CNY
TCIAB1056-1G
2,3-Bis(4-methoxyphenyl)-5-phenyltetrazolium chloride ≥98.0% (通过HPLC,滴定分析)
2,3-Bis(4-methoxyphenyl)-5-phenyltetrazolium chloride
分子式: C₂₁H₁₉ClN₄O₂
MDL编号: MFCD00060012
CAS号: 104497-79-2

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Methanol (50vol%) within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR