4-n-Octyloxyphenyl-4-butylbenzoate ≥98.0% (通过GC) liquid crystal

供应商: TCI EUROPE

TCIAB1092-1GEA 0 CNY
TCIAB1092-1G
4-n-Octyloxyphenyl-4-butylbenzoate ≥98.0% (通过GC) liquid crystal
4-n-Octyloxyphenyl-4-butylbenzoate
分子式: C₂₅H₃₄O₃
分子量: 382,54 g/mol
MDL编号: MFCD00060084
CAS号: 42815-59-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR