1,3-苯并二硫代吡咯四氟化硼盐 ≥96.0% (通过滴定分析) hydroxyl protecting agent

供应商: TCI EUROPE
Danger

Synonyms: BDTF

TCIAB1151-5GEA 0 CNY
TCIAB1151-5G
1,3-苯并二硫代吡咯四氟化硼盐 ≥96.0% (通过滴定分析) hydroxyl protecting agent
1,3-苯并二硫代吡咯四氟化硼盐
分子式: C₇H₅BF₄S₂
分子量: 240,04 g/mol
MDL编号: MFCD00070613
CAS号: 57842-27-0
UN号: 1759
ADR: 8,III

现在订购

产品技术规格

Appearance Orange to Brown to Dark purple powder to crystal
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR