1-Butoxy-4-nitrobenzene ≥99.0% (通过GC)

供应商: TCI EUROPE

TCIAB1162-25GEA 0 CNY
TCIAB1162-25G
1-Butoxy-4-nitrobenzene ≥99.0% (通过GC)
1-Butoxy-4-nitrobenzene
分子式: C₁₀H₁₃NO₃
分子量: 195,22 g/mol
MDL编号: MFCD00024688
CAS号: 7244-78-2

现在订购

产品技术规格

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 99.0 %
Freezing point 30.0 to 32.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR