2-Butyl-3-methylpyrazine ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1173-1MLEA 1350 CNY
TCIAB1173-1ML
2-Butyl-3-methylpyrazine ≥98.0% (通过GC)
2-Butyl-3-methylpyrazine
分子式: C₉H₁₄N₂
分子量: 150,23 g/mol
沸点: 84°C
MDL编号: MFCD00038031
CAS号: 15987-00-5
EINECS: 240-126-5

现在订购

产品技术规格

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR