Bis(trimethylenedithio)tetrathiafulvalene ≥97.0% (通过HPLC) for organic electronics

供应商: TCI EUROPE

TCIAB1244-100MGEA 0 CNY
TCIAB1244-100MG
Bis(trimethylenedithio)tetrathiafulvalene ≥97.0% (通过HPLC) for organic electronics
Bis(trimethylenedithio)tetrathiafulvalene
分子式: C₁₂H₁₂S₈
分子量: 412,71 g/mol
溶点: 250°C
MDL编号: MFCD00060080
CAS号: 66946-49-4

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Sulfuride) 60.0 to 63.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR