Bis(benzoylmethyl)sulphide ≥99.0% (通过GC)

供应商: TCI EUROPE

TCIAB1265-5GEA 0 CNY
TCIAB1265-5G
Bis(benzoylmethyl)sulphide ≥99.0% (通过GC)
Bis(benzoylmethyl)sulphide
分子式: C₁₆H₁₄O₂S
分子量: 270,35 g/mol
MDL编号: MFCD00054497
CAS号: 2461-80-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR