2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium chloride ≥90.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAB1311-100MGEA
0
CNY
TCIAB1311-100MG
2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium chloride ≥90.0% (通过滴定分析)
2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium chloride
分子式:
C₂₂H₁₈ClN₅O₂ |
MDL编号:
MFCD00060131 CAS号: 151390-91-9 |
产品技术规格
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...