6-Bromo-2,3-dicyanonaphthalene ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAB1390-1GEA 0 CNY
TCIAB1390-1G
6-Bromo-2,3-dicyanonaphthalene ≥98.0% (通过GC)
6-Bromo-2,3-dicyanonaphthalene
分子式: C₁₂H₅BrN₂
分子量: 257,09 g/mol
MDL编号: MFCD00130113
CAS号: 70484-02-5
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Melting point 250.0 to 254.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR