2,2-Bis(3,4-dimethylphenyl)hexafluoropropane ≥97.0% (通过GC)

TCIAB1402-5G
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane ≥97.0% (通过GC)
2,2-Bis(3,4-dimethylphenyl)hexafluoropropane
分子式: C₁₉H₁₈F₆
分子量: 360,34 g/mol
MDL编号: MFCD00042167
CAS号: 65294-20-4
EINECS: 265-687-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR