2-Benzyl-1,3-dioxolane ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB1404-25MLEA 0 CNY
TCIAB1404-25ML
2-Benzyl-1,3-dioxolane ≥98.0% (通过GC)
2-Benzyl-1,3-dioxolane
分子式: C₁₀H₁₂O₂
分子量: 164,20 g/mol
沸点: 120°C
闪点: 108°C
MDL编号: MFCD00003215
CAS号: 101-49-5
EINECS: 202-946-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR