β-histine methanesulfonate ≥99.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAB1424-25GEA 630 CNY
TCIAB1424-25G
β-histine methanesulfonate ≥99.0% (通过HPLC,滴定分析)
Betahistine methanesulfonate
分子式: C₈H₁₂N₂
MDL编号: MFCD00143688
CAS号: 54856-23-4
EINECS: 259-377-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR