1-Benzyloxy-2-methoxy-4-(1-propenyl)benzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1441-25GEA 825 CNY
TCIAB1441-25G
1-Benzyloxy-2-methoxy-4-(1-propenyl)benzene ≥98.0% (通过GC)
1-Benzyloxy-2-methoxy-4-(1-propenyl)benzene
分子式: C₁₇H₁₈O₂
分子量: 254,33 g/mol
MDL编号: MFCD00026985
CAS号: 120-11-6
EINECS: 204-370-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 61.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR