1-Bromo-2-methylhexadecane ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1456-1GEA 0 CNY
TCIAB1456-1G
1-Bromo-2-methylhexadecane ≥98.0% (通过GC)
1-Bromo-2-methylhexadecane
分子式: C₁₇H₃₅Br
分子量: 319,37 g/mol
MDL编号: MFCD00142588
CAS号: 81367-59-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR