3-tert-Butyl-5-ethyltoluene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1459-10MLEA 0 CNY
TCIAB1459-10ML
3-tert-Butyl-5-ethyltoluene ≥98.0% (通过GC)
3-tert-Butyl-5-ethyltoluene
分子式: C₁₃H₂₀
分子量: 176,30 g/mol
沸点: 71°C
闪点: 66°C
MDL编号: MFCD00026954
CAS号: 6630-01-9
EINECS: 654-362-5

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR