2,2-双(4-羟基-3,5-二甲基苯基)丙烷 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 四甲基双酚 A

TCIAB1464-25GEA 655 CNY
TCIAB1464-25G
2,2-双(4-羟基-3,5-二甲基苯基)丙烷 ≥98.0% (通过GC)
2,2-双(4-羟基-3,5-二甲基苯基)丙烷
分子式: C₁₉H₂₄O₂
分子量: 284,40 g/mol
MDL编号: MFCD00068238
CAS号: 5613-46-7
EINECS: 227-033-5

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 170.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR