Bis(tetrabutylammonium) tetracyanodiphenoquinodimethanide ≥98.0% (以总氮计)

供应商: TCI EUROPE
Warning

TCIAB1466-100MGEA 1050 CNY
TCIAB1466-100MG
Bis(tetrabutylammonium) tetracyanodiphenoquinodimethanide ≥98.0% (以总氮计)
Bis(tetrabutylammonium) tetracyanodiphenoquinodimethanide
分子式: C₅₀H₈₀N₆
MDL编号: MFCD00213933
CAS号: 68271-98-7

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystaline
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR