1,4-Bisbenzil ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAB1480-5GEA 855 CNY
TCIAB1480-5G
1,4-Bisbenzil ≥97.0% (通过GC)
1,4-Bisbenzil
分子式: C₂₂H₁₄O₄
MDL编号: MFCD00039561
CAS号: 3363-97-1

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 124.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR