1,4-Bis(3-methoxyphenoxy)benzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1486-5GEA 0 CNY
TCIAB1486-5G
1,4-Bis(3-methoxyphenoxy)benzene ≥98.0% (通过GC)
1,4-Bis(3-methoxyphenoxy)benzene
分子式: C₂₀H₁₈O₄
分子量: 322,36 g/mol
MDL编号: MFCD00142544
CAS号: 5024-84-0

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR