1,10-Bis(4-carboxyphenoxy)decane ≥97.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAB1499-5GEA 2225 CNY
TCIAB1499-5G
1,10-Bis(4-carboxyphenoxy)decane ≥97.0% (通过滴定分析)
1,10-Bis(4-carboxyphenoxy)decane
分子式: C₂₄H₃₀O₆
分子量: 414,50 g/mol
溶点: 274°C
MDL编号: MFCD00142356
CAS号: 74774-61-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization back titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR