1,4-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1512-25GEA 0 CNY
TCIAB1512-25G
1,4-Bis(4,5-dihydro-2-oxazolyl)benzene ≥98.0% (通过GC)
1,4-Bis(4,5-dihydro-2-oxazolyl)benzene
分子式: C₁₂H₁₂N₂O₂
分子量: 216,24 g/mol
MDL编号: MFCD00142545
CAS号: 7426-75-7
EINECS: 231-066-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR