(S)-(-)-1-苄基-3-吡咯烷醇 ≥98.0% (通过GC)

Warning

TCIAB1533-25GEA 2200 CNY
TCIAB1533-25G TCIAB1533-5G
(S)-(-)-1-苄基-3-吡咯烷醇 ≥98.0% (通过GC)
(S)-(-)-1-苄基-3-吡咯烷醇
分子式: C₁₁H₁₅NO
分子量: 177,25 g/mol
沸点: 120°C
MDL编号: MFCD00075485
CAS号: 101385-90-4

现在订购

产品技术规格

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Specific rotation [a]20/D +4.4 to +4.8 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR