1-Benzyl-3-(ethylamino)pyrrolidine ≥96.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAB1560-5GEA 900 CNY
TCIAB1560-5G
1-Benzyl-3-(ethylamino)pyrrolidine ≥96.0% (通过GC)
1-Benzyl-3-(ethylamino)pyrrolidine
分子式: C₁₃H₂₀N₂
MDL编号: MFCD00191305
CAS号: 115445-21-1
UN号: 2735
ADR: 8,III

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR