硼烷吡啶络合物 ≥80.0% (通过滴定分析)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
吡啶硼烷
TCIAB1569-25MLEA
565
CNY
TCIAB1569-25ML
硼烷吡啶络合物 ≥80.0% (通过滴定分析)
硼烷吡啶络合物
分子式:
C₅H₈BN 分子量: 92,94 g/mol 闪点: 21°C 存储条件: Refrigerator |
MDL编号:
MFCD00012435 CAS号: 110-51-0 EINECS: 203-773-4 UN号: 1992 ADR: 3,II |
产品技术规格
Appearance | Colorless to Brown clear liquid to slightly cloudy liquid |
Purity (Potassium iodate Method) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...