2,3-Bis(hydroxymethyl)naphthalene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1589-1GEA 0 CNY
TCIAB1589-1G
2,3-Bis(hydroxymethyl)naphthalene ≥98.0% (通过GC)
2,3-Bis(hydroxymethyl)naphthalene
分子式: C₁₂H₁₂O₂
分子量: 188,23 g/mol
MDL编号: MFCD00191314
CAS号: 31554-15-1

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 161.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR