Boc-Thr-OH ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

Synonyms: Boc-L-苏氨酸N-(叔丁氧羰基)-L-苏氨酸

TCIAB1638-10GEA 0 CNY
TCIAB1638-10G
Boc-Thr-OH ≥98.0% (通过滴定分析)
Boc-Thr-OH
分子式: C₉H₁₇NO₅
分子量: 219,2 g/mol
CAS号: 2592-18-9
EINECS: 219-987-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -7.0 to -10.0 deg (C=1, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR