4,5-双(甲硫代)-1,3-二硫杂环戊烯-2-硫酮 ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1778-1GEA 0 CNY
TCIAB1778-1G
4,5-双(甲硫代)-1,3-二硫杂环戊烯-2-硫酮 ≥98.0% (通过GC)
4,5-双(甲硫代)-1,3-二硫杂环戊烯-2-硫酮
分子式: C₅H₆S₅
分子量: 226,40 g/mol
MDL编号: MFCD00137888
CAS号: 49638-64-4

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 104.0 deg-C
Solubility in hot EtOH within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR