2-Butylbenzofuran ≥99.0% (通过GC)
供应商: TCI EUROPE
TCIAB1824-25GEA
0
CNY
TCIAB1824-25G
TCIAB1824-100G
2-Butylbenzofuran ≥99.0% (通过GC)
2-Butylbenzofuran
分子式:
C₁₂H₁₄O 分子量: 174,24 g/mol 沸点: 129°C 闪点: 101°C |
MDL编号:
MFCD00005851 CAS号: 4265-27-4 EINECS: 224-250-7 |
产品技术规格
Appearance | Light yellow to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...