1,8-Bis(hydroxymethyl)anthracene ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAB1904-1GEA
0
CNY
TCIAB1904-1G
1,8-Bis(hydroxymethyl)anthracene ≥98.0% (通过GC)
1,8-Bis(hydroxymethyl)anthracene
分子式:
C₁₆H₁₄O₂ 分子量: 238,29 g/mol |
MDL编号:
MFCD01321137 CAS号: 34824-20-9 |
产品技术规格
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 219.0 to 224.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...