Bicyclo[3.2.1]oct-2-ene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB1914-1GEA 0 CNY
TCIAB1914-1G
Bicyclo[3.2.1]oct-2-ene ≥98.0% (通过GC)
Bicyclo[3.2.1]oct-2-ene
分子式: C₈H₁₂
沸点: 135°C
MDL编号: MFCD00078236
CAS号: 823-02-9

现在订购

产品技术规格

Appearance White or Colorless powder to lump
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR