4-tert-Butyl-4'-iodobiphenyl ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB1946-5GEA 0 CNY
TCIAB1946-5G
4-tert-Butyl-4'-iodobiphenyl ≥98.0% (通过GC)
4-tert-Butyl-4'-iodobiphenyl

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 151.0 deg-C
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR