1,4-二乙酰氧基-2-丁炔 ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAB2087-25GEA 0 CNY
TCIAB2087-25G
1,4-二乙酰氧基-2-丁炔 ≥95.0% (通过GC)
1,4-二乙酰氧基-2-丁炔
分子式: C₈H₁₀O₄
分子量: 170,16 g/mol
沸点: 235°C
MDL编号: MFCD00014982
CAS号: 1573-17-7
EINECS: 216-394-4

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Freezing point 23.0 to 26.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR