3,5-Bis(4-nitrophenoxy)benzoic acid ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAB2120-5GEA 0 CNY
TCIAB2120-5G
3,5-Bis(4-nitrophenoxy)benzoic acid ≥98.0% (通过HPLC,滴定分析)
3,5-Bis(4-nitrophenoxy)benzoic acid
分子式: C₁₉H₁₂N₂O₈
分子量: 396,31 g/mol
MDL编号: MFCD02093450
CAS号: 173550-33-9

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 229.0 to 233.0 deg-C
Solubility in hot DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR