(S)-4-苄氧甲基-2,2-二甲基-1,3-二氧戊环 ≥98.0% (通过GC)

供应商: TCI EUROPE

Synonyms: 3-苄基-1,2-异亚丙基-sn-甘油D-α,β-异亚丙基甘油-γ-苄醚

TCIAB2137-1GEA 445 CNY
TCIAB2137-1G
(S)-4-苄氧甲基-2,2-二甲基-1,3-二氧戊环 ≥98.0% (通过GC)
(S)-4-苄氧甲基-2,2-二甲基-1,3-二氧戊环
分子式: C₁₃H₁₈O₃
分子量: 222,28 g/mol
闪点: 100°C
存储条件: Refrigerator
MDL编号: MFCD00043000
CAS号: 16495-03-7

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +18.0 to +21.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR