4,5-Bis(2-cyanoethylthio)-1,3-dithiole-2-thione ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Danger

TCIAB2228-1GEA 0 CNY
TCIAB2228-1G
4,5-Bis(2-cyanoethylthio)-1,3-dithiole-2-thione ≥98.0% (通过HPLC,总氮)
4,5-Bis(2-cyanoethylthio)-1,3-dithiole-2-thione
分子式: C₉H₈N₂S₅
MDL编号: MFCD01096160
CAS号: 132765-35-6
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR