Bis(ethylenediamine)palladium(II) dichloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAB2244-1GEA
0
CNY
TCIAB2244-1G
Bis(ethylenediamine)palladium(II) dichloride ≥98.0% (通过滴定分析)
Bis(ethylenediamine)palladium(II) dichloride
分子式:
C₄H₁₆Cl₂N₄Pd 分子量: 297,52 g/mol |
MDL编号:
MFCD03844778 CAS号: 16483-18-4 EINECS: 240-538-5 |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...