N-苄基吡咯 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAB2335-5GEA
0
CNY
TCIAB2335-5G
N-苄基吡咯 ≥98.0% (通过GC)
N-苄基吡咯
分子式:
C₁₁H₁₁N 分子量: 157,22 g/mol 沸点: 245°C 溶点: 15°C 闪点: 46°C |
MDL编号:
MFCD00963301 CAS号: 2051-97-0 EINECS: 218-137-1 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...