2-Bromobenzyl phenyl sulphone ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB2367-1GEA 0 CNY
TCIAB2367-1G
2-Bromobenzyl phenyl sulphone ≥98.0% (通过GC)
2-Bromobenzyl phenyl sulphone
分子式: C₁₃H₁₁BrO₂S
分子量: 311,19 g/mol
MDL编号: MFCD06797069
CAS号: 92022-50-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR