2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAB2618-1GEA 0 CNY
TCIAB2618-1G
2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene ≥97.0% (通过GC)
2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene
分子式: C₁₀H₁₄O₄
分子量: 198,22 g/mol
CAS号: 78840-04-7

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR