(1R,2R,4R,5R)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAB2652-100MGEA
0
CNY
TCIAB2652-100MG
(1R,2R,4R,5R)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane ≥98.0% (通过HPLC)
(1R,2R,4R,5R)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane
分子式:
C₃₇H₅₄N₂O₂ 分子量: 558,85 g/mol |
CAS号:
539834-19-0 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...