1-Bromo-3,5-bis(tert-butylthio)benzene ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAB2658-1GEA
0
CNY
TCIAB2658-1G
1-Bromo-3,5-bis(tert-butylthio)benzene ≥98.0% (通过GC)
1-Bromo-3,5-bis(tert-butylthio)benzene
分子式:
C₁₄H₂₁BrS₂ 分子量: 333,35 g/mol |
CAS号:
795274-44-1 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...