3,5-Bis(tert-butylthio)benzyl bromide ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAB2660-1GEA 0 CNY
TCIAB2660-1G
3,5-Bis(tert-butylthio)benzyl bromide ≥98.0% (通过GC)
3,5-Bis(tert-butylthio)benzyl bromide
分子式: C₁₅H₂₃BrS₂
分子量: 347,37 g/mol
存储条件: Refrigerator
UN号: 3261
ADR: 8,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR