9,9-Bis(4-amino-3-fluorophenyl)fluorene ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAB2691-5GEA 0 CNY
TCIAB2691-5G
9,9-Bis(4-amino-3-fluorophenyl)fluorene ≥98.0% (通过HPLC,总氮)
9,9-Bis(4-amino-3-fluorophenyl)fluorene
分子式: C₂₅H₁₈F₂N₂
分子量: 384,43 g/mol
MDL编号: MFCD08276304
CAS号: 127926-65-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR